missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ARID1B (aa 915-1030) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53438 (PA5-53438. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This locus encodes an AT-rich DNA interacting domain-containing protein. The encoded protein is a component of the SWI/SNF chromatin remodeling complex and may play a role in cell-cycle activation. The protein encoded by this locus is similar to AT-rich interactive domain-containing protein 1A. These two proteins function as alternative, mutually exclusive ARID-subunits of the SWI/SNF complex. The associated complexes play opposing roles. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq, Feb 2012].
Specifications
Specifications
| Accession Number | Q8NFD5 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57492 |
| Name | Human ARID1B (aa 915-1030) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 6A3-5; 8030481M12; 9330189K18Rik; AI836955; Ardi1b; ARID domain-containing protein 1 B; ARID1B; AT rich interactive domain 1 B (Swi1 like); AT rich interactive domain 1 B (SWI1-like); AT rich interactive domain 1 B (SWI-like); AT-rich interaction domain 1 B; AT-rich interactive domain-containing protein 1 B; B230217J03Rik; BAF250B; BRG1-associated factor 250 b; BRG1-binding protein ELD/OSA1; BRG1-binding protein hELD/OSA1; BRIGHT; CSS1; DAN15; ELD (eyelid)/OSA protein; ELD/OSA1; hOsa2; KIAA1235; mKIAA1235; MRD12; Osa homolog 2; OSA2; P250R; Smcf1; smcf-1; transcription factor 1 |
| Common Name | ARID1B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction