Learn More
Abnova™ Human ARHGEF1 Full-length ORF (NP_004697.2, 1 a.a. - 912 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009138-P02.25ug
Additional Details : Weight : 0.00010kg
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. [provided by RefSeq]
Sequence: MEDFARGAASPGPSRPGLVPVSIIGAEDEDFENELETNSEEQNSQFQSLEQVKRRPAHLMALLQHVALQFEPGPLLCCLHADMLGSLGPKEAKKAFLDFYHSFLEKTAVLRVPVPPNVAFELDRTRADLISEDVQRRFVQEVVQSQQVAVGRQLEDFRSKRLMGMTPWEQELAQLEAWVGRDRASYEARERHVAERLLMHLEEMQHTISTDEEKSAAVVNAIGLYMRHLGVRTKSGDKKSGRNFFRKKVMGNRRSDEPAKTKKGLSSILDAARWNRGEPQVPDFRHLKAEVDAEKPGATDRKGGVGMPSRDRNIGAPGQDTPGVSLHPLSLDSPDREPGADAPLELGDSSPQGPMSLESLAPPESTDEGAETESPEPGDEGEPGRSGLELEPEEPPGWRELVPPDTLHSLPKSQVKRQEVISELLVTEAAHVRMLRVLHDLFFQPMAECLFFPLEELQNIFPSLDELIEVHSLFLDRLMKRRQESGYLIEEIGDVLLARFDGAEGSWFQKISSRFCSRQSFALEQLKAKQRKDPRFCAFVQEAESRPRCRRLQLKDMIPTEMQRLTKYPLLLQSIGQNTEEPTEREKVELAAECCREILHHVNQAVRDMEDLLRLKDYQRRLDLSHLRQSSDPMLSEFKNLDITKKKLVHEGPLTWRVTKDKAVEVHVLLLDDLLLLLQRQDERLLLKSHSRTLTPTPDGKTMLRPVLRLTSAMTREVATDHKAFYVLFTWDQEAQIYELVAQTVSERKNWCALITETAGSLKVPAPASRPKPRPSPSSTREPLLSSSENGNGGRETSPADARTERILSDLLPFCRPGPEGQLAATALRKVLSLKQLLFPAEEDNGAGPPRDGDGVPGGGPLSPARTQEIQENLLSLEETMKQLEELEEEFCRLRPLLSQLGGNSVPQPGCTSpecifications
NP_004697.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
128.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ARHGEF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9138 | |
ARHGEF1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GEF1/LBCL2/LSC/P115-RHOGEF/SUB1.5 | |
ARHGEF1 | |
Recombinant | |
wheat germ expression system |