missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ARHGAP32 (aa 1076-1170) Control Fragment Recombinant Protein

Product Code. 30209776
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209776

Brand: Invitrogen™ RP102828

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58365 (PA5-58365. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

GTPase-activating protein (GAP) promoting GTP hydrolysis on RHOA, CDC42 and RAC1 small GTPases. May be involved in the differentiation of neuronal cells during the formation of neurite extensions. Involved in NMDA receptor activity-dependent actin reorganization in dendritic spines. May mediate cross-talks between Ras- and Rho regulated signaling pathways in cell growth regulation. Isoform 2 has higher GAP activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A7KAX9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9743
Name Human ARHGAP32 (aa 1076-1170) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3426406O18Rik; 6430596G11Rik; AI841135; ARHGAP32; Arhgap35; brain-specific Rho GTP-ase-activating protein; brain-specific Rho GTPase-activating protein; BRF2; butyrate response factor 2; EGF-response factor 2; ERF2; ERF-2; GAB-associated CDC42; GAB-associated Cdc42/Rac GTPase-activating protein; GAP-associated protein p190; Gc-gap; glucocorticoid receptor DNA binding factor 1; glucocorticoid receptor DNA-binding factor 1; Glucocorticoid receptor repression factor 1; GRF1; GRF-1; Grit; Grlf1; GTPase regulator interacting with TrkA; GTPase-activating protein for Cdc42 and Rac1; KIAA0712; KIAA1722; mKIAA0712; mKIAA1722; mRNA decay activator protein ZFP36L2; P190 RhoGAP; p190A; p190-A; p190ARHOGAP; p190RhoGAP; p200RhoGAP; p250Gap; PX-RICS; rac GTPase activating protein; rho GAP p190A; Rho GTPase activating protein 32; Rho GTPase activating protein 33 pseudogene; Rho GTPase activating protein 35; rho GTPase-activating protein 32; Rho GTPase-activating protein 35; Rho/Cdc42/Rac GTPase-activating protein RICS; rhoGAP involved in the beta-catenin-N-cadherin and NMDA receptor signaling; RhoGAP involved in the -catenin-N-cadherin and NMDA receptor signaling; rho-type GTPase-activating protein 32; RICS; RNF162C; TIS11D; TPA-induced sequence 11 d; ZFP36 ring finger protein like 2; ZFP36 ring finger protein-like 2; Zfp36l2; ZFP36-like 2; zinc finger protein 36, C3H type-like 1; zinc finger protein 36, C3H type-like 2; zinc finger protein 36, C3H type-like 2-like; zinc finger protein 36, C3H1 type-like 2; zinc finger protein, C3H type, 36-like 2
Common Name ARHGAP32
Gene Symbol ARHGAP32
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AVATTEDNLSSSYSAVALDKAYFQTDRPAEQFHLQNNAPGNCDHPLPETTATGDPTHSNTTESGEQHHQVDLTGNQPHQAYLSGDPEKARITSVP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.