missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human APRIL Recombinant Protein, PeproTech®

Product Code. 30131761
Click to view available options
Quantity:
10 x 50 μg
20 x 50 μg
Unit Size:
Pack of 2
Pack of 4
This item is not returnable. View return policy

Product Code. 30131761

Brand: Gibco™ 31010C500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

310-10C-01M will be provided as 20 x 50 μg (310-10C-50UG). 310-10C-500 will be provided as 10 x 50 μg (310-10C-50UG). 310-10C-250 will be provided as 5 x 50 μg (310-10C-50UG). 310-10C-100 will be provided as 2 x 50 μg (310-10C-50UG) Recombinant human APRIL is a soluble 16.3 kDa protein, consisting of 146 amino acid residues. *Human APRIL (Catalog Number 310-10C) has replaced Human APRIL (Catalog Number 310-10). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75888
For Use With (Application) Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 8741
Molecular Weight (g/mol) 16.3 kDa
Name Human APRIL
Purification Method Purified
Quantity 10 x 50 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 2310026N09Rik; A proliferation-inducing ligand; APRIL; CD256; TALL2; TALL-2; TNF- and APOL-related leukocyte expressed ligand 2; TNF-related death ligand 1; Tnfsf13; TNLG7B; Trdl1; TRDL-1; tumor necrosis factor (ligand) superfamily, member 13; tumor necrosis factor ligand 7B; tumor necrosis factor ligand superfamily member 13; tumor necrosis factor superfamily member 13; tumor necrosis factor-like protein ZTNF2; tumor necrosis factor-related death ligand-1; UNQ383/PRO715; ZTNF2
Common Name APRIL
Gene Symbol TNFSF13
Biological Activity Determined by its ability to induce NF-kappaB activation in HEK293-NF-kappaB reporter cells transfected with BCMA receptor.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence AVLTQKQKKQ HSVLHLVPIN ATSKDDSDVT EVMWQPALRR GRGLQAQGYG VRIQDAGVYL LYSQVLFQDV TFTMGQVVSR EGQGRQETLF RCIRSMPSHP DRAYNSCYSA GVFHLHQGDI LSVIIPRARA KLNLSPHGTF LGFVKL
Content And Storage -20°C
Activity Determined by its ability to induce NF-kappaB activation in HEK293-NF-kappaB reporter cells transfected with BCMA receptor.
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.