missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APPL Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7075
This item is not returnable.
View return policy
Description
APPL Monoclonal specifically detects APPL in Human samples. It is validated for Western Blot.
Specifications
| APPL | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UKG1 | |
| APPL1 | |
| E. coli-derived recombinant human APPL Met1-Ile373 Accession # Q9UKG1 | |
| 100 μg | |
| Primary | |
| Detects human APPL in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 709208 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Adapter protein containing PH domain, PTB domain and leucine zipper motif 1, adaptor protein containing pH domain, PTB domain and leucine zipper motif 1, adaptor protein, phosphotyrosine interaction, PH domain and leucine zippercontaining 1, AKT2 interactor, APPL1, APPLdip13-alpha, DCC-interacting protein 13-alpha, DIP 13 alpha, DIP13A, Dip13-alpha, DIP13alpha, KIAA1428, signaling adaptor protein DIP13alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26060 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction