missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APP/Protease Nexin II (Research Grade Solanezumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
£179.00 - £370.00
Specifications
| Antigen | APP |
|---|---|
| Clone | Hu13 |
| Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
| Applications | CyTOF, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18190251
|
R&D Systems
MAB9919-SP |
25 μg |
£179.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18149230
![]() |
R&D Systems
MAB9919-100 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APP Monoclonal specifically detects APP in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| APP | |
| Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II | |
| Human APP/Protease Nexin II | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects Human APP/Protease Nexin II based on Solanezumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Solanezumab. This product is for research use only. |
| Hu13 | |
| CyTOF, Flow Cytometry | |
| Unconjugated | |
| Human | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| 351 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title