missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APP/Protease Nexin II Pan Specific Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1168
This item is not returnable.
View return policy
Description
APP Polyclonal specifically detects APP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| APP | |
| Polyclonal | |
| Unconjugated | |
| NP_958817 | |
| APP | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP 695 +1 frameshift mutant Leu18-Arg336 Accession # NP_958817 | |
| 100 μg | |
| Primary | |
| Detects human APP695 +1 and human APP770 in direct ELISAs and Western blots. This antibody recognizes a common epitope present on APP695 +1 and APP770 found within the segments Leu18 - Arg288 or Pro365 - Arg411 of APP770 (Accession P05067). | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 351 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction