missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Apolipoprotein O (aa 133-198) Control Fragment Recombinant Protein

Product Code. 30209491
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30209491 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30209491 Supplier Invitrogen™ Supplier No. RP92263

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51853 (PA5-51853. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of the apolipoprotein family. Members of this protein family are involved in the transport and metabolism of lipids. The encoded protein associates with HDL, LDL and VLDL lipoproteins and is characterized by chondroitin-sulfate glycosylation. This protein may be involved in preventing lipid accumulation in the myocardium in obese and diabetic patients. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 3, 4, 5, 12 and 16.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BUR5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79135
Name Human Apolipoprotein O (aa 133-198) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610008C08Rik; 1110019O03Rik; Apolipoprotein O; APOO; brain my025; FAM121B; family with sequence similarity 121 B; MGC4825; Mic23; MIC26; MICOS complex subunit Mic23; MICOS complex subunit MIC26; My025; MYO25; Protein FAM121B; RGD1565289; UNQ1866/PRO4302
Common Name Apolipoprotein O
Gene Symbol APOO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IKKLVYPPGFMGLAASLYYPQQAIVFAQVSGERLYDWGLRGYIVIEDLWKENFQKPGNVKNSPGTK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.