missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Human APOL1 ELISA Kit

Product Code. 16841466 Shop All Proteintech Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Type Unit Size
16841466 Plasma, Serum 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16841466 Supplier Proteintech™ Supplier No. KE0004796T
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous APOL1

  • Recognizes human APOL1
  • Human apolipo-protein L1 (APOL1) is a minor component of plasma high density lipoprotein (HDL) particles
    • Thought to be predominantly involved in lipid transport and metabolism
    • Involved in host innate immunity against Trypanosoma parasites
      • Once activated, can lyse parasite and protect human from infection
    • Variants in APOL1 gene associated with chronic kidney disease (FSGS, HIVAN) and hypertensive nephropathy

Process:

  • Antibody specific for APOL1 pre-coated onto microwells
  • APOL1 protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody specific for APOL 1 is added to detect captured APOL1 protein
  • For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number NP_001130012
Assay Range 0.156 to 10 ng/mL
Assay Sensitivity 0.07 ng/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 8542
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID Q2KHQ6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.