missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human ApoE3 Recombinant Protein, PeproTech®

Product Code. 30088565 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
500 μg
Unit Size:
100µg
500µg
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30088565 2 x 500 μg Pack of 2
17802613 100 μg 100µg
17852043 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30088565 Supplier Gibco™ Supplier No. 3500201M
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

350-02-1MG will be provided as 2 x 500 μg (350-02-500UG). Recombinant Human ApoE3 is a 34.4 kDa protein containing 300 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

ApoE belongs to a group of proteins that bind reversibly with lipoprotein and play an important role in lipid metabolism. In addition to facilitating solubilization of lipids, these proteins help to maintain the structural integrity of lipoproteins, serve as ligands for lipoprotein receptors, and regulate the activity of enzymes involved in lipid metabolism. Significant quantities of ApoE are produced in the liver and brain, and to some extent in almost every organ. ApoE is an important constituent of all plasma lipoproteins. Its interaction with specific ApoE receptor enables uptake of chylomicron remnants by liver cells, which is an essential step during normal lipid metabolism. It also binds with the LDL receptor (apo B/E). Defects in ApoE are a cause of hyperlipoproteinemia type III. ApoE exists in three major isoforms; E2, E3, and E4, which differ from one another by a single amino-acid substitution. E3 is the most common isoform and is present in 40-90% of the population.
TRUSTED_SUSTAINABILITY

Especificaciones

Accession Number P02649
For Use With (Application) Bioactivity, Control, Western Blot Control
Formulation Protein with no preservative
Gene ID (Entrez) 348
Molecular Weight (g/mol) 34.4 kDa
Name Human ApoE3
Purification Method Purified
Quantity 2 x 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias ApoE
Common Name ApoE3
Gene Symbol APOE
Biological Activity Determined by its ability to stimulate the proliferation of human SH-SY5Y cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKVEQAVETE PEPELRQQTE WQSGQRWELA LGRFWDYLRW VQTLSEQVQE ELLSSQVTQE LRALMDETMK ELKAYKSELE EQLTPVAEET RARLSKELQA AQARLGADME DVCGRLVQYR GEVQAMLGQS TEELRVRLAS HLRKLRKRLL RDADDLQKRL AVYQAGAREG AERGLSAIRE RLGPLVEQGR VRAATVGSLA GQPLQERAQA WGERLRARME EMGSRTRDRL DEVKEQVAEV RAKLEEQAQQ IRLQAEAFQA RLKSWFEPLV EDMQRQWAGL VEKVQAAVGT SAAPVPSDNH
Content And Storage -20°C
Activity Determined by its ability to stimulate the proliferation of human SH-SY5Y cells.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.