missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOC3 native Protein from human plasma

Product Code. 16292100
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16292100 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16292100 Supplier Abnova™ Supplier No. P4280.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for SDS-PAGE

Apolipoprotein C-III is a very low density lipoprotein (VLDL) protein. APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia. [provided by RefSeq]

Specifications

For Use With (Application) SDS-PAGE
Formulation In 10mM NH
Gene ID (Entrez) 345
Molecular Weight (g/mol) 8.75kDa
Name APOC3 (Human) Native Protein
Preparation Method Native protein purified from human plasma.
Quantity 100 μg
Storage Requirements Store at -80°C. After reconstitution with 100 ul reagent grade water to generate 1 mg/mL solution, store at -80°C. Aliquot to avoid repeated freezing and thawing. Allow the protein solution to sit for 10 minutes with gentle agitation prior to use.
Regulatory Status RUO
Gene Alias APOCIII/MGC150353
Common Name APOC3
Gene Symbol APOC3
Species Human
Protein Tag None
Expression System Native protein purified from human plasma
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.