missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human APOBEC3B (aa 255-279) Control Fragment Recombinant Protein

Product Code. 30211339
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211339

Brand: Invitrogen™ RP105404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (52%), Rat (52%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66863 (PA5-66863. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. A hybrid gene results from the deletion of approximately 29.5 kb of sequence between this gene, APOBEC3B, and the adjacent gene APOBEC3A. The breakpoints of the deletion are within the two genes, so the deletion allele is predicted to have the promoter and coding region of APOBEC3A, but the 3' UTR of APOBEC3B. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UH17
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9582
Name Human APOBEC3B (aa 255-279) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias A3B; A3Z2_1; A3Z2-Z3; A3Z3; A3Z3_1; APOBEC1L; Apobec3; Apobec3b; Apobec3f; APOBEC3F1; APOBEC3F2; APOBEC3Z2; APOBEC3Z3; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3 B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3 B; apolipoprotein B mRNA editing enzyme catalytic subunit 3 B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3 B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3 F; apolipoprotein B mRNA-editing complex 3; ARCD3; Arp3; ARP4; BC003314; bK150C 2.2; bK150C2.2; Cem15; CEM-15; cytidine deaminase; DJ742C19.2; DNA dC->dU-editing enzyme APOBEC3; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3 B; FLJ21201; phorbolin 2; phorbolin 3; phorbolin-1-related protein; Phorbolin-2/3; PHRBNL; probable DNA dC->dU-editing enzyme APOBEC3; probable DNA dC->dU-editing enzyme APOBEC-3 B; recovery from Friend virus 3; Rfv3
Common Name APOBEC3B
Gene Symbol APOBEC3B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ELRFLDLVPSLQLDPAQIYRVTWFI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.