missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APLP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4945
This item is not returnable.
View return policy
Description
APLP-2 Polyclonal specifically detects APLP-2 in Human samples. It is validated for Western Blot.
Specifications
| APLP-2 | |
| Polyclonal | |
| Unconjugated | |
| AAH00373 | |
| APLP2 | |
| Mouse myeloma cell line NS0-derived recombinant human APLP-2 Leu37-Ser680 Accession # AAH00373 | |
| 100 μg | |
| Primary | |
| Detects human APLP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse (rm) APLP-2 is observed and less than 1% cross-reactivity with recombinant human APLP-1 and rmAPLP-1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP2, APLP-2, APPH, APPHCDEI box-binding protein, APPL2, CDEBP, WALPLP2, YWK-II | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 334 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction