missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APLP-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3129
This item is not returnable.
View return policy
Description
APLP-1 Polyclonal specifically detects APLP-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.
Specifications
| APLP-1 | |
| Polyclonal | |
| Unconjugated | |
| P51693 | |
| APLP1 | |
| Mouse myeloma cell line NS0-derived recombinant human APLP-1 Gln34-Glu580 Accession # P51693 | |
| 100 μg | |
| Primary | |
| Detects human APLP-1 in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse APLP-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 25 ug/mL, Immunohistochemistry 0.3-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 333 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction