missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human APLP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £370.00
Specifications
| Antigen | APLP-1 |
|---|---|
| Clone | 354008 |
| Concentration | LYOPH |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18760938
![]() |
R&D Systems
MAB3129-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18133311
|
R&D Systems
MAB3129 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APLP-1 Monoclonal specifically detects APLP-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| APLP-1 | |
| LYOPH | |
| Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P51693 | |
| 333 | |
| Mouse myeloma cell line NS0-derived recombinant human APLP-1 Gln34-Glu580 (predicted) Accession # P51693 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human APLP-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse APLP-1 is observed and no cross-reactivity with recombinant human APLP-2 is observed. |
| 354008 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1 | |
| APLP1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title