missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human APC6 Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
APC6 Synthetic Peptide, PEP-1061, from Invitrogen.
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
Specifications
Specifications
| Gene ID (Entrez) | 8881 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q13042 |
| Common Name | Human APC6 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction