missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human APC15 (aa 6-69) Control Fragment Recombinant Protein

Product Code. 30212277
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212277

Brand: Invitrogen™ RP97313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57736 (PA5-57736. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ANAPC15 is a component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. In the complex, it plays a role in the release of the mitotic checkpoint complex (MCC) from the APC/C: not required for APC/C activity itself, but promotes the turnover of CDC20 and MCC on the APC/C, thereby participating to the responsiveness of the spindle assembly checkpoint. It is also required for degradation of CDC20.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60006
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25906
Name Human APC15 (aa 6-69) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3200002M19Rik; 6330414C15Rik; Anapc15; anaphase promoting complex subunit 15; anaphase prompoting complex C subunit 15; anaphase-promoting complex subunit 15; APC15; C11orf51; C15H11orf51; C21H11orf51; HSPC020; hypothetical protein LOC505356; protein HSPC020 homolog; RGD1311634; uncharacterized protein C11orf51 homolog; Unknown (protein for MGC:153425); zgc:153425
Common Name APC15
Gene Symbol Anapc15
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSLFPRVTETLWFNLDRPCVEETELQQQEQQHQAWLQSIAEKDNNLVPIGKPASEHYDDEEEED
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.