missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AP3S2 (aa 107-154) Control Fragment Recombinant Protein

Product Code. 30181371
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181371

Brand: Invitrogen™ RP99268

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61921 (PA5-61921. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

AP3S2 (Adaptor Related Protein Complex 3 Subunit Sigma 2) is a Protein Coding gene. Among its related pathways are Lysosome. Gene Ontology (GO) annotations related to this gene include protein transporter activity. An important paralog of this gene is AP3S1. Part of the AP-3 complex, an adaptor-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. In concert with the BLOC-1 complex, AP-3 is required to target cargos into vesicles assembled at cell bodies for delivery into neurites and nerve terminals.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P59780
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10239
Name Human AP3S2 (aa 107-154) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias [s]3 B; adapter-related protein complex 3 sigma-2 subunit; adapter-related protein complex 3 subunit sigma-2; adaptor complex sigma3B; adaptor related protein complex 3 sigma 2 subunit; adaptor related protein complex 3, sigma 2 subunit; adaptor-related protein complex 3 subunit sigma-2; adaptor-related protein complex 3, sigma 2 subunit; adaptor-related protein complex AP-3, sigma 2 subunit; AP-3 complex sigma-3 B subunit; AP-3 complex subunit sigma-2; AP-3 complex subunit sigma-3 B; AP3S2; AP3S3; clathrin-associated/assembly/adaptor protein, small 4, 22-kD; FLJ35955; sigma 3 B; sigma3b; sigma-3 B-adaptin; sigma3B-adaptin; Sigma-adaptin 3 b
Common Name AP3S2
Gene Symbol AP3S2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LDLIFHMDKVHYILQEVVMGGMVLETNMNEIVAQIEAQNRLEKSEGGL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.