missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AP3M1 Full-length ORF (NP_036227.1, 1 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16198442
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16198442 25 μg 25µg
16188442 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16198442 Supplier Abnova™ Supplier No. H00026985P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is the medium subunit of AP-3, which is an adaptor-related protein complex associated with the Golgi region as well as more peripheral intracellular structures. AP-3 facilitates the budding of vesicles from the Golgi membrane and may be directly involved in protein sorting to the endosomal/lysosomal system. AP-3 is a heterotetrameric protein complex composed of two large subunits (delta and beta3), a medium subunit (mu3), and a small subunit (sigma 3). Mutations in one of the large subunits of AP-3 have been associated with the Hermansky-Pudlak syndrome, a genetic disorder characterized by defective lysosome-related organelles. Alternatively spliced transcript variants encoding the same protein have been observed. [provided by RefSeq]

Sequence: MIHSLFLINCSGDIFLEKHWKSVVSQSVCDYFFEAQEKAADVENVPPVISTPHHYLISIYRDKLFFVSVIQTEVPPLFVIEFLHRVADTFQDYFGECSEAAIKDNVVIVYELLEEMLDNGFPLATESNILKELIKPPTILRSVVNSITGSSNVGDTLPTGQLSNIPWRRAGVKYTNNEAYFDVVEEIDAIIDKSGSTVFAEIQGVIDACIKLSGMPDLSLSFMNPRLLDDVSFHPCIRFKRWESERVLSFIPPDGNFRLISYRVSSQNLVAIPVYVKHSISFKENSSCGRFDITIGPKQNMGKTIEGITVTVHMPKVVLNMNLTPTQGSYTFDPVTKVLTWDVGKITPQKLPSLKGLVNLQSGAPKPEENPSLNIQFKIQQLAISGLKVNRLDMYGEKYKPFKGVKYVTKAGKFQVRT

Specifications

Accession Number NP_036227.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26985
Molecular Weight (g/mol) 73.3kDa
Name AP3M1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC22164
Common Name AP3M1
Gene Symbol AP3M1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.