missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AP-2 epsilon Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5060-SP
This item is not returnable.
View return policy
Description
AP-2 epsilon/TFAP2E Polyclonal specifically detects AP-2 epsilon/TFAP2E in Human samples. It is validated for Western Blot.
Specifications
| AP-2 epsilon/TFAP2E | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activating enhancer-binding protein 2-epsilon, adaptor-related protein complex 2, epsilon subunit, AP2 epsilon, AP2E, AP2-epsilon, MGC49007, TFAP2E, transcription factor AP-2 epsilon (activating enhancer binding protein 2epsilon), transcription factor AP-2-epsilon | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 339488 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6VUC0 | |
| TFAP2E | |
| E. coli-derived recombinant human AP-2 epsilon Ala123-Val217 Accession # Q6VUC0 | |
| 25 μg | |
| Primary | |
| Detects human AP-2 epsilon in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) AP-2 alpha , rhAP-2 beta , rhAP-2 delta , and rhAP-2 gamma is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction