missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ANPEP (967 amino acids) native Protein from human serum

Product Code. 16058076
Change view
Click to view available options
Quantity:
10 ug
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16058076 10 ug 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16058076 Supplier Abnova™ Supplier No. P4918.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Aminopeptidase N is located in the small-intestinal and renal microvillar membrane, and also in other plasma membranes. In the small intestine aminopeptidase N plays a role in the final digestion of peptides generated from hydrolysis of proteins by gastric and pancreatic proteases. Its function in proximal tubular epithelial cells and other cell types is less clear. The large extracellular carboxyterminal domain contains a pentapeptide consensus sequence characteristic of members of the zinc-binding metalloproteinase superfamily. Sequence comparisons with known enzymes of this class showed that CD13 and aminopeptidase N are identical. The latter enzyme was thought to be involved in the metabolism of regulatory peptides by diverse cell types, including small intestinal and renal tubular epithelial cells, macrophages, granulocytes, and synaptic membranes from the CNS. Human aminopeptidase N is a receptor for one strain of human coronavirus that is an important cause of upper respiratory tract infections. Defects in this gene appear to be a cause of various types of leukemia or lymphoma. [provided by RefSeq]

Specifications

For Use With (Application) Functional Study, SDS-PAGE
Formulation In 20mM Tris-HCl, 500mM NaCl, 1mM CaCl
Gene ID (Entrez) 290
Molecular Weight (g/mol) 110-130kDa
Name ANPEP (Human) Native Protein
Preparation Method Native protein purified from human serum
Purification Method Saltprecipitation/Ion Exchange Chromatography/Gelfiltration
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 10 ug
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias APN/CD13/LAP1/PEPN/gp150/p150
Common Name ANPEP
Gene Symbol ANPEP
Biological Activity > 500mU/mg (L-Leu-pNA at 37°C, pH 7.2).
Species Human
Protein Tag None
Expression System Native protein purified from human serum
Form Liquid
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.