missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ANO7 (aa 23-95) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (37%), Rat (37%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57378 (PA5-57378. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Has calcium-dependent phospholipid scramblase activity; scrambles phosphatidylserine, phosphatidylcholine and galactosylceramide. Does not exhibit calcium-activated chloride channel (CaCC) activity. May play a role in cell-cell interactions.
Specifications
Specifications
| Accession Number | Q6IWH7 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 50636 |
| Name | Human ANO7 (aa 23-95) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | ANO7; anoctamin 7; anoctamin-7; Dresden transmembrane protein of the prostate; Dresden-transmembrane protein of the prostate; DTMPP; D-TMPP; IPCA5; IPCA-5; New gene expressed in prostate; NGEP; PCANAP5; PCANAP5L; prostate cancer associated protein 5; Prostate cancer-associated protein 5; TMEM16G; Transmembrane protein 16 G |
| Common Name | ANO7 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur