missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Annexin A8/ANXA8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8105
This item is not returnable.
View return policy
Description
Annexin A8/ANXA8 Polyclonal specifically detects Annexin A8/ANXA8 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Annexin A8/ANXA8 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| annexin A8, Annexin VIII, Annexin-8, Anx8, ANXA8, ANXA8L2, FLJ53095, VAC-beta, Vascular anticoagulant-beta | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 653145 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P13928 | |
| ANXA8 | |
| E. coli-derived recombinant human Annexin A8/ANXA8 Ala2-Pro327 Accession # P13928 | |
| 100 μg | |
| Primary | |
| Detects human Annexin A8/ANXA8 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) ANXA4, rhANXA5, rhANXA6, rhANXA7, rhANXA11, and rhANXA13A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction