missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Annexin A1 ELISA Kit (Colorimetric)

Product Code. 18678440 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18678440

Brand: Novus Biologicals™ NBP260538

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.313 to 20 ng/mL (example only, lot dependent)
Assay Sensitivity 0.29 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Culture
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 301
Gene Symbol ANXA1
Quantity 1 Kit
Research Discipline Protein Kinase, Signal Transduction
Storage Requirements Storage is content dependent.
Synonym annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.