missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ANKMY1 (aa 369-499) Control Fragment Recombinant Protein

Product Code. 30202287
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202287

Brand: Invitrogen™ RP102334

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (20%), Rat (20%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55359 (PA5-55359. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ANKMY1 (ankyrin repeat and MYND domain containing 1), also known as ZMYND13 or TSAL1, is a 941 amino acid protein that contains seven ANK repeats, three MORN repeats and one MYND-type zinc finger. MORN repeats were first identified in junctophilins, cytoplasmic proteins involved in junctions between the plasma membrane and the ER/SR membrane. The presence of MORN repeats suggests that ANKMY1 may interact with the plasma membrane. The MYND domain consists of a cluster of cysteine and histidine residues, arranged with an invariant spacing to form a potential zinc-binding motif which may be involved in protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9P2S6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51281
Name Human ANKMY1 (aa 369-499) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias ANKMY1; ankyrin repeat and MYND domain containing 1; ankyrin repeat and MYND domain-containing protein 1; Testis-specific ankyrin-like protein 1; TSAL1; Zinc finger MYND domain-containing protein 13; ZMYND13
Common Name ANKMY1
Gene Symbol ANKMY1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SSFMDTNLESLYYEVNVPSQGSYELRPPPAPLLLPRVSGSHEGGHFQDTGQCGGSIDHRSSSLKGDSPLVKGSLGHVESGLEDVLGNTDRGSLCSAETKFESNVCVCDFSIELSQAMLERSAQSHSLLKMA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.