missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human ANGPTL-3 Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
Description
Recombinant human ANGPTL-3 is a glycoprotein that migrates by SDS-PAGE analysis at an apparent molecular weight of 62 kDa, and contains 452 amino acid residues including a C-terminal His tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This gene encodes a member of a family of secreted proteins that function in angiogenesis. The encoded protein, which is expressed predominantly in the liver, is further processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metabolism, while the C-terminal chain may be involved in angiogenesis. Mutations in this gene cause familial hypobetalipoproteinemia type 2.
Specifications
Specifications
| Accession Number | Q9Y5C1 |
| For Use With (Application) | Control, ELISA Standard, Binding Assay, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 27329 |
| Molecular Weight (g/mol) | 52.9 kDa |
| Name | Human ANGPTL-3 |
| Purification Method | Purified |
| Quantity | 100 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction