missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiostatin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF226-SP
This item is not returnable.
View return policy
Description
Angiostatin Polyclonal specifically detects Angiostatin in Human samples. It is validated for Western Blot.
Specifications
| Angiostatin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PLG | |
| E. coli-derived recombinant human Angiostatin Kringle 1-3 Val98-Ser358 with Cys188Ser & Cys316Ser substitutions Accession # P00747 | |
| 25 μg | |
| Primary | |
| Detects human Angiostatin in direct ELISAs and Western blots. This antibody will also recognize human plasminogen. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P00747 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5340 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction