missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-like Protein 6/ANGPTL6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£169.00 - £522.00
Specifications
| Antigen | Angiopoietin-like Protein 6/ANGPTL6 |
|---|---|
| Clone | 856606 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18703507
![]() |
R&D Systems
MAB11306-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18733667
![]() |
R&D Systems
MAB11306-100 |
100 μg |
£522.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-like Protein 6/ANGPTL6 Monoclonal antibody specifically detects Angiopoietin-like Protein 6/ANGPTL6 in Human samples. It is validated for ELISASpecifications
| Angiopoietin-like Protein 6/ANGPTL6 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| AGF, AGFAngiopoietin-related protein 5, Angiopoietin like Protein 6, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ANGPTL6, ARP5, ARP5angiopoietin-related protein 6 | |
| E. coli-expressed recombinant Human Angiopoietin-like Protein 6/ANGPTL6, Arg21-Leu470, Accession # Q8ROZ6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 856606 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8ROZ6 | |
| 83854 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title