missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-like Protein 4/ANGPTL4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£196.00 - £460.00
Specifications
| Antigen | Angiopoietin-like Protein 4/ANGPTL4 |
|---|---|
| Clone | 944740 |
| Dilution | 0.25 μg/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18725785
|
R&D Systems
MAB11305-100 |
100 μg |
£460.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795885
|
R&D Systems
MAB11305-SP |
25 μg |
£196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human samples. It is validated for Flow CytometrySpecifications
| Angiopoietin-like Protein 4/ANGPTL4 | |
| 0.25 μg/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| angiopoietin-like 4, Angiopoietin-like protein 4, ARP4fasting-induced adipose factor, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| Mouse myeloma cell line, NS0-derived human Angiopoietin-like Protein 4/ANGPTL4 protein, Gly26-Ser406, Accession # Q9BY76 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 944740 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BY76 | |
| 51129 | |
| IgG2b | |
| Protein A or G purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title