missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Angiopoietin-like 6, Carrier-Free Recombinant Protein

Product Code. 18716296
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18716296

Brand: R&D Systems™ 8466AN025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human Angiopoietin-like Protein 6/ANGPTL6 (Catalog # 8466-AN/CF) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human LILRB2/CD85d/ILT4 Fc Chimera (Catalog # 2078-T4 ) binds with an ED50 of 30.0-360 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NI99
For Use With (Application) Bioactivity
Gene ID (Entrez) 83854
Molecular Weight (g/mol) Observed M.W.: 58-69kDa and 31-41kDa, reducing conditions, Theoretical M.W.: 51kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 6/ANGPTL6 protein Ala20-Leu470,with an N-terminal c-Myc tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias AGF, AGFAngiopoietin-related protein 5, Angiopoietin like Protein 6, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ANGPTL6, ARP5, ARP5angiopoietin-related protein 6
Common Name Angiopoietin-like Protein 6/ANGPTL6
Gene Symbol ANGPTL6
Biological Activity Measured by its ability to promote the expansion of E16 rat liver mononuclear cells in vitro, in the presence of Recombinant Mouse SCF/c-kit Ligand, Recombinant Mouse Thrombopoietin/Tpo, and Recombinant Mouse Flt-3 Ligand. The ED50 for this effect is typically 8-40ng/mL in the presence of a cross-linking Mouse Anti-Human c-Myc Monoclonal Antibody.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ala20-Leu470, with an N-terminal c-Myc tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.