missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-2 Biotinylated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems BAM0981
This item is not returnable.
View return policy
Description
Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).
Specifications
| Angiopoietin-2 | |
| Monoclonal | |
| LYOPH | |
| ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| O15123 | |
| ANGPT2 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-2 Asp68-Phe496 Accession # O15123 | |
| 250 μg | |
| Primary | |
| Detects human Angiopoietin-2 in ELISAs. In ELISAs, this antibody shows no cross-reactivity with recombinant human (rh) Ang-1, rhAng-4, rhAng-X, rmAng-3, rmANGPTL3, rhTie-1, rhTie-2, and rmTie-2. | |
| Human | |
| Purified |
| ELISA | |
| 85834 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, ANGPT2, Tie2-ligand | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 285 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction