missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Angiopoietin-1 Quantikine ELISA Kit

Product Code. 18782044
Change view
Click to view available options
Quantity:
1 Plate
6 Plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782044 1 Plate 1 set
18712354 6 Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782044 Supplier R&D Systems™ Supplier No. DANG10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Angiopoietin-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Angiopoietin-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Angiopoietin-1 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), platelet-poor citrate plasma (10 ul), saliva (25 ul), human milk (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.50 - 4000pg/mL
Assay Sensitivity 10.3pg/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Platelet-poor Plasma (Citrate), Saliva, Serum
Specificity Natural and Recombinant Human Angiopoietin-1
Sufficient For Sufficient materials to run ELISAs on at least one 96-well plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 80381
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Platelet-poor Citrate Plasma (10uL),Saliva (25uL),Human Milk (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target B7-H3/CD276
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.