missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Human Angiopoietin 1 ELISA Kit

Product Code. 17234876 Shop All Proteintech Products
Change view
Click to view available options
Gene ID (Entrez):
284
Unit Size:
96 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gene ID (Entrez) Unit Size
17234876 284 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17234876 Supplier Proteintech Supplier No. KE0021296T
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Angiopoietin 1 is a 70-kDa secreted glycoprotein generated from vascular mural cells, pericytes, and certain other cells.

Angiopoietin 1 participates in vascular differentiation through angiogenesis, which is the process of the growth and remodelling of existing vessels. Angiopoietin 1 is also involved in the maintenance and turnover of blood vessels in mature animals. Angiopoietin1 binds to and activates the TEK/TIE2 receptor by inducing its dimerization and tyrosine phosphorylation, playing an important role in the regulation of angiogenesis. Overexpression of Angiopoietin1 has been proven to occur in malignant glioblastoma, neuroblastoma, non-small cell lung cancer and other tumors.

Specifications

Gene ID (Entrez) 284
compliance-icons compliance-icons
Product Identifier
  • 96RXN Human Angiopoietin 1 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.