missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems BAF923
This item is not returnable.
View return policy
Description
Angiopoietin-1 Polyclonal specifically detects Angiopoietin-1 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Angiopoietin-1 | |
| Polyclonal | |
| Biotin | |
| Q5HYA0 | |
| ANGPT1 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-1 (R&D Systems, Catalog # 923-AN) Ser20-Phe498 Accession # Q5HYA0 | |
| 50 μg | |
| Primary | |
| Detects human Angiopoietin-1 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) Angiopoietin-2, rmAngiopoietin-3, and rhAngiopoietin-4 is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, ANGPT1, KIAA0003 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 284 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction