Learn More
Human Angiopoietin-1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 7 publications
Brand:  R&D Systems MAB923-SP
 View more versions of this product
Product Code. 18770068
  • £170.00 / 25µg
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Angiopoietin-1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, ANGPT1, KIAA0003
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
284
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
Western Blot, Immunohistochemistry
171718
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Q15389
ANGPT1
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-1 Ser20-Phe498 Accession # Q15389
25 μg
Primary
Detects human Angiopoietin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Angiopoietin-2, recombinant mouse Angiopoietin-like 3, rhAngiopoietin-4, or rhAngiopoietin-like 7 is observed.
Human
Purified