missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human ANG-1 Recombinant Protein, PeproTech®

Product Code. 17891033
Click to view available options
Quantity:
10 x 100 μg
100 μg
20 μg
5 μg
5 x 100 μg
Unit Size:
100µg
1mg
20µg
500µg
5µg
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 17891033

Brand: Gibco™ 130061MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

130-06-1MG will be provided as 10 x 100 μg (130-06-100UG). 130-06-500UG will be provided as 5 x 100 μg (130-06-100UG). Recombinant Human ANG-1, derived from HeLa cells, is a C-terminal histidine tagged glycoprotein which migrates with an apparent molecular mass of 60.0-70.0 kDa by SDS-PAGE under reducing conditions. Sequencing analysis shows N-terminal sequences starting with Ser-20 and with Asp-70 of the 498 amino acid precursor protein. The calculated molecular weight of Recombinant Human ANG-1 is 56.3 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15389
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 284
Molecular Weight (g/mol) 56.3
Name Human ANG-1
Quantity 10 x 100 μg
Source HeLa cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 1110046O21Rik; 2810039D03Rik; AGP1; Agpt; Agpt1; ANG1; ang-1; ANG3; ANG-3; angioarrestin; angiopoietin 1; angiopoietin 3; angiopoietin like 1; angiopoietin Y1; angiopoietin-1; Angiopoietin-3; angiopoietin-like 1; angiopoietin-like protein 1; Angiopoietin-related protein 1; Angpt1; ANGPT3; Angptl1; AngY; ARP1; dJ595C2.2; dJ595C2.2 (angiopoietin Y1); endothelial growth factor; KIAA0003; PSEC0154; tie2 receptor ligand; UNQ162; UNQ162/PRO188
Gene Symbol Angpt1
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Sequence SNQRRSPENS GRRYNRIQHG QCAYTFILPE HDGNCRESTT DQYNTNALQR DAPHVEPDFS SQKLQHLEHV MENYTQWLQK LENYIVENMK SEMAQIQQNA VQNHTATMLE IGTSLLSQTA EQTRKLTDVE TQVLNQTSRL EIQLLENSLS TYKLEKQLLQ QTNEILKIHE KNSLLEHKIL EMEGKHKEEL DTLKEEKENL QGLVTRQTYI IQELEKQLNR ATTNNSVLQK QQLELMDTVH NLVNLCTKEG VLLKGGKREE EKPFRDCADV YQAGFNKSGI YTIYINNMPE PKKVFCNMDV NGGGWTVIQH REDGSLDFQR GWKEYKMGFG NPSGEYWLGN EFIFAITSQR QYMLRIELMD WEGNRAYSQY DRFHIGNEKQ NYRLYLKGHT GTAGKQSSLI LHGADFSTKD ADNDNCMCKC ALMLTGGWWF DACGPSNLNG MFYTAGQNHG KLNGIKWHYF KGPSYSLRST TMMIRPLDFH HHHHH
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.