missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Androgen R/NR3C4 Isoform 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £388.00
Specifications
| Antigen | Androgen R/NR3C4 |
|---|---|
| Clone | 1022602 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792857
![]() |
R&D Systems
MAB58764-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721037
![]() |
R&D Systems
MAB58764-100 |
100 μg |
£388.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for ImmunocytochemistrySpecifications
| Androgen R/NR3C4 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AIS, androgen receptor, AndrogenR, AR, DHTR, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Human Androgen R/NR3C4 isoform 2 synthetic peptide, Accession # P10275 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1022602 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P10275 | |
| 367 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title