missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AMPKβ2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£188.00 - £467.00
Specifications
| Antigen | AMPK beta 2/PRKAB2 |
|---|---|
| Clone | 1092615 |
| Dilution | Western Blot 2 μg/mL, Simple Western 20 μg/mL |
| Applications | Western Blot, Simple Western |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30281240
|
R&D Systems
MAB11639-100 |
100 μg |
£467.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30281241
|
R&D Systems
MAB11639-SP |
25 μg |
£188.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AMPK beta 2/PRKAB2 Monoclonal antibody specifically detects AMPK beta 2/PRKAB2 in Human samples. It is validated for Western Blot,Simple WesternSpecifications
| AMPK beta 2/PRKAB2 | |
| Western Blot 2 μg/mL, Simple Western 20 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, PRKAB2, protein kinase, AMP-activated, beta 2 non-catalytic subunit | |
| E. coli - derived recombinant human AMPKbeta2, Met1-Ile272, Accession # O43741 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1092615 | |
| Western Blot, Simple Western | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43741 | |
| 5565 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title