missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Amphiregulin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems BAF262
This item is not returnable.
View return policy
Description
Amphiregulin Polyclonal specifically detects Amphiregulin in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Amphiregulin | |
| Polyclonal | |
| Biotin | |
| P15514 | |
| AREG | |
| E. coli-derived recombinant human Amphiregulin Val107-Lys184 Accession # P15514 | |
| 50 μg | |
| Primary | |
| Detects human Amphiregulin in ELISAs and Western blots. In sandwich immunoassays, less than 0.05% cross-reactivity with rmAR, rhEGF and rhTGF-alpha is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 374 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction