missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Amphiregulin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB262-SP
This item is not returnable.
View return policy
Description
Amphiregulin Monoclonal specifically detects Amphiregulin in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Amphiregulin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AREG | |
| E. coli-derived recombinant human Amphiregulin | |
| 25 μg | |
| Primary | |
| Detects human Amphiregulin (AR) in Western blots. In this format, this antibody shows less than 5% cross-reactivity with recombinant human HB-EGF. | |
| Human | |
| Purified |
| Western Blot, Neutralization, ELISA | |
| 31221 | |
| Western Blot 1 ug/mL, Neutralization 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 374 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction