missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Amphiphysin/AMPH Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6527-SP
This item is not returnable.
View return policy
Description
Amphiphysin/AMPH Monoclonal specifically detects Amphiphysin/AMPH in Human samples. It is validated for Immunohistochemistry.
Specifications
| Amphiphysin/AMPH | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P49418 | |
| AMPH | |
| E. coli-derived recombinant human Amphiphysin/AMPH Asn204-Gly300 Accession # P49418 | |
| 25 μg | |
| Primary | |
| Detects human Amphiphysin/AMPH in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 694145 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AMPH, AMPH1, amphiphysin, amphiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen), amphiphysin (Stiff-Mann syndrome with breast cancer 128kD autoantigen), amphiphysin I | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 273 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction