missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Amnionless Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1860
This item is not returnable.
View return policy
Description
Amnionless/AMN Polyclonal specifically detects Amnionless/AMN in Human samples. It is validated for Western Blot.
Specifications
| Amnionless/AMN | |
| Polyclonal | |
| Unconjugated | |
| Q9BXJ7 | |
| AMN | |
| Mouse myeloma cell line NS0-derived recombinant human Amnionless Val20-Ala358 Accession # Q9BXJ7 | |
| 100 μg | |
| Primary | |
| Detects human Amnionless in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AMN, amnionless homolog (mouse), PRO1028, protein amnionless, UNQ513/PRO1028, visceral endoderm-specific type 1 transmembrane protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 81693 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction