missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aminopeptidase P1/XPNPEP1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF2970
Additional Details : Weight : 0.00970kg
Description
Aminopeptidase P1/XPNPEP1 Polyclonal specifically detects Aminopeptidase P1/XPNPEP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Aminopeptidase P1/XPNPEP1 | |
Polyclonal | |
Unconjugated | |
NP_065116 | |
XPNPEP1 | |
E. coli-derived recombinant human Aminopeptidase P1/XPNPEP1 Met1-His623 Accession # NP_065116 | |
100 μg | |
Primary | |
Detects human Aminopeptidase P1/XPNPEP1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) XPNPEP2 is observed. | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Aminoacylproline aminopeptidase, APP1, cytosolic, Cytosolic aminopeptidase P, EC 3.4.11.9, sAmp, xaa-Pro aminopeptidase 1, XPNPEP1, XPNPEPLX-prolyl aminopeptidase (aminopeptidase P)-like, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, X-prolyl aminopeptidase 1, soluble | |
Goat | |
Affinity Purified | |
RUO | |
7511 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only