missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aminopeptidase LRAP/ERAP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£170.00 - £403.00
Specifications
| Antigen | Aminopeptidase LRAP/ERAP2 |
|---|---|
| Clone | 3F5 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783529
![]() |
R&D Systems
MAB3830-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18171631
![]() |
R&D Systems
MAB3830 |
100 μg |
£403.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase LRAP/ERAP2 Monoclonal specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blot.Specifications
| Aminopeptidase LRAP/ERAP2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6P179 | |
| 64167 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Aminopeptidase LRAP/ERAP2 Accession # Q6P179 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Aminopeptidase LRAP/ERAP2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human PILS is observed. |
| 3F5 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, ERAP2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.- | |
| ERAP2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title