missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AMICA/JAML Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB34491S-100UG
This item is not returnable.
View return policy
Description
AMICA/JAML Monoclonal specifically detects AMICA/JAML in Human samples. It is validated for Flow Cytometry.
Specifications
| AMICA/JAML | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q86YT9 | |
| AMICA1 | |
| Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML Leu20-Leu275 Accession # Q86YT9 | |
| RUO | |
| 120425 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 401901 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human AMICA/JAML in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse AMICA is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction