missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AMICA (aa 297-379) Control Fragment Recombinant Protein

Product Code. 30206555
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206555

Brand: Invitrogen™ RP88832

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (42%), Rat (42%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61621 (PA5-61621. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

AMICA transmembrane protein of the plasma membrane of leukocytes that control their migration and activation through interaction with CXADR, a plasma membrane receptor found on adjacent epithelial and endothelial cells. The interaction between both receptors mediates the activation of gamma-delta T-cells, a subpopulation of T-cells residing in epithelia and involved in tissue homeostasis and repair. Upon epithelial CXADR-binding, JAML induces downstream cell signaling events in gamma-delta T-cells through PI3-kinase and MAP kinases. It results in proliferation and production of cytokines and growth factors by T-cells that in turn stimulate epithelial tissues repair. It also controls the transmigration of leukocytes within epithelial and endothelial tissues through adhesive interactions with epithelial and endothelial CXADR.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86YT9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 120425
Name Human AMICA (aa 297-379) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias adhesion molecule AMICA; adhesion molecule interacting with CXADR antigen 1; adhesion molecule, interacts with CXADR antigen 1; AMICA; Amica1; Crea7; CREA7-1; CREA7-4; Dendritic cell-specific protein CREA7; Dendritic cell-specific protein CREA7-1; dendritic-cell specific protein Crea7; dendritic-cell specific protein CREA7-1; dendritic-cell specific protein CREA7-4; Gm638; Jaml; junction adhesion molecule like; junctional adhesion molecule-like; mCrea7; UNQ722/PRO1387
Common Name AMICA
Gene Symbol JAML
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KKTCGNKSSVNSTVLVKNTKKTNPEIKEKPCHFERCEGEKHIYSPIIVREVIEEEEPSEKSEATYMTMHPVWPSLRSDRNNSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.