missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £512.00
Specifications
| Antigen | alpha-Synuclein |
|---|---|
| Clone | 1060506 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725845
![]() |
R&D Systems
MAB11376-100 |
100 μg |
£512.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755955
![]() |
R&D Systems
MAB11376-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| alpha-Synuclein | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha, truncated alpha synuclein | |
| E. coli -derived human alpha-Synuclein, Met1-Ala140, Accession # P37840 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1060506 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P37840 | |
| 6622 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title