missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Synuclein Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1338
This item is not returnable.
View return policy
Description
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human samples. It is validated for Western Blot.
Specifications
| alpha-Synuclein | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P37840 | |
| SNCA | |
| E. coli-derived recombinant human alpha-Synuclein Met1-Ala140 Accession # P37840 | |
| 500 μg | |
| Primary | |
| Detects human alpha-Synuclein in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 189105 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha, truncated alpha synuclein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6622 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction