missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-N-acetylgalactosaminidase/NAGA Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6717-SP
This item is not returnable.
View return policy
Description
alpha-N-acetylgalactosaminidase/NAGA Polyclonal specifically detects alpha-N-acetylgalactosaminidase/NAGA in Human samples. It is validated for Western Blot.
Specifications
| alpha-N-acetylgalactosaminidase/NAGA | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-N- (alpha-galactosidase B), alphaNacetylgalactosaminidase, alpha-N-acetylgalactosaminidase, EC 3.2.1, EC 3.2.1.49, GALB, N-acetylgalactosaminidase, alpha-, NAGA | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4668 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P17050 | |
| NAGA | |
| Chinese hamster ovary cell line recombinant human alpha-N-acetylgalactosaminidase/NAGA Leu18-Gln411 Accession # P17050 | |
| 25 μg | |
| Primary | |
| Detects human alpha-N-acetylgalactosaminidase/NAGA in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human alpha-Galactosidase A is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction