missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha-Internexin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4844-SP
This item is not returnable.
View return policy
Description
alpha-Internexin Monoclonal specifically detects alpha-Internexin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| alpha-Internexin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q16352 | |
| INA | |
| E. coli-derived recombinant human alpha-Internexin Val230-Glu450 Accession # Q16352 | |
| 25 μg | |
| Primary | |
| Detects human alpha-Internexin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) NF-M or rhVimentin is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 529724 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 66 kDa neurofilament protein, alphaInternexin, alpha-Inx, FLJ18662, FLJ57501, INA, internexin neuronal intermediate filament protein, alpha, MGC12702, NEF5, Neurofilament 5, neurofilament 5 (66kD), Neurofilament-66, neurofilament-66, tax-binding protein, NF-66, NF-66alpha-internexin, TXBP-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9118 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction